![Nitrendipine for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/398/512/959f6d12-f354-41a3-a8f3-15de0f1be7bb/640/959f6d12-f354-41a3-a8f3-15de0f1be7bb.png)
Nitrendipine for peak identification
![Nitrendipine for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/398/512/959f6d12-f354-41a3-a8f3-15de0f1be7bb/640/959f6d12-f354-41a3-a8f3-15de0f1be7bb.png)
Synonym(s):
Nitrendipine, 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid ethyl methyl ester
Empirical Formula (Hill Notation):
C18H20N2O6
CAS Number:
Molecular Weight:
360.36
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing