
Lysostaphin from Staphylococcus staphylolyticus

Synonym(s):
Glycyl-glycine Endopeptidase, Lysostaphin from Staphylococcus staphylolyticus
CAS Number:
Products
Product Category
Brand
Biological Source
Physical Form
Product Line
Purity
Activity Spectrum
Chemical Composition
Quality Segment
Application
Mode of Action
Sterilization Method
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing