
Ketorolac Related Compound C

Synonym(s):
5-Benzoyl-2,3-dihydro-1H-pyrrolizin-1-one
Empirical Formula (Hill Notation):
C14H11NO2
CAS Number:
Molecular Weight:
225.24
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing