
Gemcitabine impurity A

Synonym(s):
Cytosine, 4-Amino-2-hydroxypyrimidine, 4-Aminopyrimidin-2-(1H)-one
Empirical Formula (Hill Notation):
C4H5N3O
CAS Number:
Molecular Weight:
111.10
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing