
(±)-Phenylpropanolamine hydrochloride solution

Synonym(s):
DL-Norephedrine hydrochloride, (±)-Phenylpropanolamine hydrochloride, erythro-2-Amino-1-phenyl-1-propanol hydrochloride, DL-erythro-2-Amino-1-phenyl-1-propanol hydrochloride, DL-Phenylpropanolamine hydrochloride
Linear Formula:
C6H5CH(OH)CH(NH2)CH3 · HCl
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing