
(±)-Mevalonolactone-(methyl-d3)

Synonym(s):
Mevalonolactone-(methyl-d3), DL-Mevalonic-methyl-d3 lactone
Empirical Formula (Hill Notation):
C6D3H7O3
Molecular Weight:
133.16
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing