![Carvedilol Related Compound E United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/324/459/e7eec32e-510f-426a-bc5c-83f4956ff1d3/640/e7eec32e-510f-426a-bc5c-83f4956ff1d3.png)
Carvedilol Related Compound E
![Carvedilol Related Compound E United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/324/459/e7eec32e-510f-426a-bc5c-83f4956ff1d3/640/e7eec32e-510f-426a-bc5c-83f4956ff1d3.png)
Synonym(s):
2-(2-Methoxyphenoxy)ethylamine
Empirical Formula (Hill Notation):
C9H13NO2
CAS Number:
Molecular Weight:
167.21
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing