
Apigenin 7-O-glucuronide

Synonym(s):
Apigenin 7-glucuronide, 4′,5,7-Trihydroxyflavone 7-glucuronide, Scutellarin
Empirical Formula (Hill Notation):
C21H18O11
CAS Number:
Molecular Weight:
446.36
Products
Formula Weight
Manufacturer
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing