
Anteiso 15:0 fatty acid

Synonym(s):
Anteiso 15:0 Fatty Acid
Empirical Formula (Hill Notation):
C15H30O2
CAS Number:
Molecular Weight:
242.40
Products
Product Category
Formula Weight
Manufacturer
Physical Form
Purity
Package Size
Showing 1-1 of 1
Compare
Product Number
Description
Pricing