Skip to Content
Merck
Adrenocorticotropic Hormone human ≥97% (HPLC)

Adrenocorticotropic Hormone human

Adrenocorticotropic Hormone human ≥97% (HPLC)
Synonym(s):
Adrenocorticotropic hormone fragment 1-39, Corticotropin A
Empirical Formula (Hill Notation):
C207H308N56O58S
CAS Number:
Molecular Weight:
4541.07

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing