
Adrenaline impurity F

Synonym(s):
(−)-Epinephrine, (−)-Adrenalin, (R)-(−)-3,4-Dihydroxy-α-(methylaminomethyl)benzyl alcohol, L-Adrenaline, L-Epinephrine
Linear Formula:
(HO)2C6H3CH(OH)CH2NHCH3
CAS Number:
Molecular Weight:
183.20
Products
Formula Weight
Manufacturer
Melting Point (°C)
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing