Skip to Content
Merck
2-Bromomethyl-anthraquinone 97%

2-Bromomethyl-anthraquinone

2-Bromomethyl-anthraquinone 97%
Synonym(s):
MAQ-Br
Empirical Formula (Hill Notation):
C15H9BrO2
CAS Number:
Molecular Weight:
301.13

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing