
2-(4-Benzoyl-3-hydroxyphenoxy)ethyl acrylate

Linear Formula:
H2C=CHCO2CH2CH2OC6H3(OH)COC6H5
CAS Number:
Molecular Weight:
312.32
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Monomer Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing