XX1614701
Millipore Glass Cover for Vacuum Filtration
47 mm, For use with Millipore MilliSolve Kit
Synonym(s):
Cover for Vacuum Filtration, Glass Cover for Filtration, Millipore Filtration Cover
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24
Recommended Products
material
borosilicate glass
manufacturer/tradename
MilliSolve
technique(s)
vacuum filtration: suitable
application(s)
filtration
shipped in
ambient
General description
Millipore Glass Vacuum Filtration Accessory Product
Glass cover, 47 mm, For use with Millipore MilliSolve Kit
These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
This item is an accessory part for the Millipore MilliSolve Kit.
Glass cover, 47 mm, For use with Millipore MilliSolve Kit
These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
This item is an accessory part for the Millipore MilliSolve Kit.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service