Automated IgG (Immunoglobulin G) Purification Using PureProteome™ Magnetic Beads With the KingFisher® Particle Processor
Introduction
As the demand for antigen-specific immunoglobulins increases in research, diagnostic and therapeutic settings, so increases the need for high reproducibility and batch-to-batch consistency of antibodies and antibody-based reagents. To meet requirements for both high throughput and low variability, many laboratories have turned to automated systems for antibody purification using protein A or protein G magnetic beads.
Compared to traditional purification methods using agarose either in batch mode or packed into spin columns, which requires cumbersome centrifugation steps, automated magnetic bead processors are able to consistently process multiple samples at once while reducing hands-on time. Moreover, the process is highly reproducible and there is no risk of cross-contamination between samples.
Liquid handling and bead handling robots are the two types of systems commonly used for automating magnetic bead separation protocols. In liquid handling robots, magnetic separation is achieved by integrating an external 96-well microtiter magnetic rack to separate the beads from the liquid. Alternatively, bead handling robots such KingFisher® particle processors use magnetic rods covered with disposable plastic tips to move beads from one solution to the next.
PureProteome™ Protein A and G Magnetic beads provide a rapid and reproducible means to purify immunoglobulins from complex mixtures such as serum, plasma or cell culture supernatant samples. For IgG purification, the samples are mixed with PureProteome™ Protein G Magnetic Beads for a short period of time to bind the immunoglobulins. The beads are then isolated by magnetic separation, followed by several wash steps to remove unbound proteins. Finally, the bound proteins are eluted at high purity for use in further downstream applications. Here, we evaluated the effectiveness of PureProteome™ Protein G Magnetic Beads for IgG purification in a fully automated purification process using the KingFisher® Duo particle processor. To obtain the best results, parameters such as mixing speed, bead collection time and duration of binding and washing steps were optimized for use with PureProteome™ Protein G Magnetic Beads.
Materials and Methods
Antibody Purification─Standard Manual Protocol
IgG purification from rabbit serum was performed using 100 μL of suspended PureProteome™ Protein G Magnetic Bead slurry (Cat. No. LSKMAGG10) in 1.5 mL microcentrifuge tubes (6 replicates). Phosphate-buffered saline (PBS) containing 0.01% Tween® 20 detergent (Cat. No. 655205) was used as binding/wash buffer. Using the PureProteome™ Magnetic Stand (Cat. No. LSKMAGS08), the beads were washed with 500 μL of binding buffer by vigorously vortexing for 10 seconds, collecting the beads on the magnet and removing the buffer with a pipette. Rabbit serum (25 μL) was diluted to 200 μL with binding buffer and added to the beads. The mixture was incubated at room temperature for 30 minutes with continuous, end–over-end mixing. The unbound fraction was discarded, and, as described above, the beads were washed three times with 500 μL of wash buffer using the PureProteome™ Magnetic Stand to capture beads. Finally, the elution of IgG was performed by adding 100 μL of 0.2 M glycine HCl, pH 2.5 to the beads and mixing at room temperature for 1–2 minutes. An additional elution was performed in a similar manner with 50 μL to achieve maximum yield. Both elutions were collected using the PureProteome™ Magnetic Stand into the same microcentrifuge tube and saved for further analysis.
Antibody Purification─KingFisher® Duo Protocol
Initially, all the reagents and samples were pipetted into the KingFisher® Duo plates (Microtiter Deepwell 96 plate and elution strips) and a protocol was set up on King-Fisher® Duo System (Thermo Fisher) according to the plate layout and optimized conditions (mixing and collecting parameters for PureProteome™ magnetic beads) outlined in Table 1. Similarly to the manual protocol, 100 μL of suspended PureProteome™ Protein G Magnetic Bead slurry was used for IgG purification using KingFisher® Duo System. PBS containing 0.01% Tween® 20 detergent was used as binding/wash buffer and 25 μL of rabbit serum diluted in a total volume of 200 μL of binding buffer was pipetted into row D of the Microtiter Deepwell 96 plate and the binding step was performed for 30 minutes using medium mixing speed. Elution was carried out with 0.2 M glycine HCl, pH 2.5 in elution strips for better recovery of the sample. The protocol was executed and the plates were loaded onto the KingFisher® Duo System. After the run was completed, the plates were removed and the purified samples were collected for further analysis.
Plate/Row or Elution Strip | Row Name | Content | Volume | Mixing Time/Speed | Collecting Count/Time |
---|---|---|---|---|---|
1-A | Beads | Protein G Beads | 100 μL | 1 min/Medium | 4/10 sec |
1-B | Tip | 12-Tip | Comb | - | - |
1-C | Equilibration | Binding Buffer | 500 μL | 1 min/Medium | 4/10 sec |
1-D | Bind | Sample + Binding Buffer | 200 μL | 30 min/Medium | 4/10 sec |
1-E | Wash 1 | Wash Buffer | 500 μL | 1 min/Medium | 4/10 sec |
1-F | Wash 2 | Wash Buffer | 500 μL | 1 min/Medium | 4/10 sec |
1-G | Wash 3 | Wash Buffer | 500 μL | 1 min/Medium | 4/10 sec |
Elution Strip 1 | Elution 1 | Elution Buffer | 100 μL | 1 min/Medium | 4/10 sec |
Elution Strip 2 | Elution 2 | Elution Buffer | 50 μL | 1 min/Medium | 4/10 sec |
Protein Quantitation by Bradford Assay
10 μL of IgG standards and samples were added to a microtiter plate followed by 300 μL of Coomassie™ reagent. The reactions were mixed briefly on a plate shaker and analyzed by detecting the absorbance of the samples at 595 nm and the concentration was determined using the IgG standard curve.
Sodium Dodecyl Sulfate Polyacrylamide Gel Electrophoresis (SDS-PAGE)
Purified samples (10 μL of the eluted fractions) and control rabbit serum (5 μL of starting material) were reduced and denatured at 70 °C for 10 min and loaded onto 1 mm thick 4–12% gradient NuPAGE® Bis-Tris gels (Life Technologies). Proteins were separated at 200 V for 35 min. After electrophoresis, gels were removed from the cassette, briefly rinsed and washed 3 times with Milli-Q® water and stained with SimplyBlue™ SafeStain using the microwave protocol. Stained gels were then washed in Milli-Q® water for 10 minutes and imaged.
Results
Protein Yield
The purified samples obtained with the automated and manual protocols were analyzed by Bradford assay to determine protein yield. The results show that the total amount of IgG eluted using the KingFisher® system is similar to that of the manual process (Table 2).
SDS-PAGE and Coomassie® Staining
The eluted fractions were also analyzed by SDS-PAGE for specificity of the elution and reproducibility. Visual inspection of the stained gel (Figure 1) demonstrate comparable purity and yield, indicating that PureProteome™ Protein G Magnetic Beads can be used to generate reproducible batches of IgGs using KingFisher® particle processor.
No. of Samples | μg IgG eluted | % CV | |
---|---|---|---|
KingFisher® System | 1 | 181.62 | 6.25 |
Manual | 6 | 195.84 | 5.22 |
Figure 1.SDS-PAGE gel: IgG purification from rabbit serum using PureProteome™ Protein G Magnetic Beads. Elution of the antibody was performed using 0.2 M glycine HCl pH 2.5. Samples were separated by SDS-PAGE and bands were visualized by Coomassie® staining (Molecular weight standard is shown in leftmost lane; SM=starting material; remaining lanes are eluted fractions from KingFisher® Duo and eluted fractions from manual testing).
Network error: Failed to fetch