Merck
HomeLysis & Protein ExtractionBugBuster® and Benzonase® Reagents are the Clear Solutions to Simple, Efficient Extraction of E. coli Proteins
BugBuster® and Benzonase® Reagents are the Clear Solutions to Simple, Efficient Extraction of E. coli Proteins
Anthony Grabski, Mark McCormick, Robert Mierendorf
Novagen, Inc.
There are a variety of methods for cellular disintegration and extraction of proteins from E. coli ranging from enzymatic digestion and osmotic shock to ultrasonication and pressure disruption. Each method has inherent advantages and disadvantages. Generally, vigorous mechanical treatments reduce viscosity but can result in heat and oxidative inactivation of labile proteins, while gentle treatments may not release the target protein from the cells, and resulting extracts are extremely viscous. In addition to proteins, bacterial extracts contain large amounts of nucleic acid, ribosomal material, and dispersed cell wall polysaccharide resulting in a viscous gum-like mass. This viscosity is problematic during the initial steps of downstream processing.
Extraction and recovery of proteins expressed in E. coli has been simplified through the application of two powerful bioprocessing reagents: BugBuster® Protein Extraction Reagent and Benzonase® Nuclease. BugBuster® reagents are formulated for the gentle disruption of the E. coli cell wall, resulting in the liberation of soluble proteins. It provides a simple, rapid, low cost alternative to mechanical disruption methods such as French press or sonication for releasing target proteins in preparation for functional studies or purification. The proprietary formulation utilizes a mixture of non-ionic detergents that are capable of cell wall perforation without denaturing soluble proteins.
The Benzonase® reagent is a genetically engineered endonuclease produced and purified from E. coli strain W3110, a mutant of strain K12, containing the proprietary pNUC1 production plasmid.1,2 The plasmid encodes an endonuclease classified as a nonspecific phosphodiesterase originally isolated from Serratia marcescens.3,4 Structurally, the protein is a dimer of identical 245 amino acid, ~30 kDa subunits with two essential disulfide bonds.5–8 This promiscuous endonuclease attacks and degrades all forms of DNA and RNA (single stranded, double stranded, linear and circular) and is effective over a wide range of operating conditions (Table 1, ref. 9). The enzyme completely digests nucleic acids to 5'-monophosphate terminated oligonucleotides 2–5 bases in length.4,10 Although the nuclease is capable of cleavage at nearly all positions along a nucleic acid chain, sequence dependent preferences have been demonstrated.11 The enzyme prefers GC-rich regions in dsDNA while avoiding d(A):d(T)-tracts. Benzonase® reagent is available in two grades, Purity > 99% and Purity > 90%. Both preparations possess exceptionally high specific activities and are supplied free from measurable protease activities and viral contaminants. Benzonase® reagent is ideal for a wide variety of applications where complete digestion of nucleic acids is desirable. This article demonstrates the combination of BugBuster® and Benzonase® reagents as protein purification tools to extract recombinant proteins from E. coli and to reduce the viscosity of the resulting extract.
Table 1Operating conditions for Benzonase® Nuclease.
Viscosity reduction by Benzonase E. coli cells
Figure 1.Viscosity reduction by Benzonase nuclease E. coli cells BL21(DE3) containing a pET construct were suspended in BugBuster® reagent (5 mL/g wet weight). Aliquots of the suspension were treated with the indicated amounts of Benzonase® nuclease for 10 min at room temperature, centrifuged at 350 × g for 3 min and photographed.
Extract preparation
The E. coli cells BL21(DE3) containing a pET construct encoding a His•Tag® fusion protein were grown in liquid culture, induced with IPTG for 5 h, harvested by centrifugation, and cell pellets frozen before BugBuster + Benzonase reagents extraction. Frozen cell pellets were completely resuspended at room temperature in BugBuster® reagent (5 mL/g wet weight) containing 0.5 mM PMSF and 15 µg/mL benzamidine. The crude suspension was divided into two equal aliquots. One aliquot was treated with 1 µL (25 units) Benzonase® nuclease (Purity > 90%) per mL of BugBuster® reagent used in the initial resuspension. The second control aliquot was not treated with Benzonase® nuclease. These crude extracts were reacted on a rotary mixer for 15–20 min at room temperature. Insoluble cell debris was removed by centrifugation at 16,000 × g for 20 min at 4 °C. The soluble supernatant fractions were carefully removed with a pipet and saved for purification of the target protein. The insoluble pellets were discarded.
Viscosity reduction
The results of Benzonase® nuclease treatment and its influence on viscosity reduction and nucleic acid digestion are clearly demonstrated in Figures. 1 and 2, respectively. Figure 1 demonstrates viscosity reduction through a simple centrifugation experiment employing 0, 2.5, and 25 units of Benzonase®/mL and BugBuster® reagents extract of E. coli cells. Benzonase® nuclease treatment significantly reduced the viscosity of crude cell extracts, allowing effective clarification with lower centrifugal force than is required to clarify untreated extract. In Figure 1, significant clarification of the extracts was seen with only 350 × g centrifugal force. Typically, untreated E. coli extracts are clarified by centrifugation at 20,000–50,000 × g in order to pellet cell wall fragments and membranes. After Benzonase® nuclease reatment, extracts can be clarified by centrifugal forces of only 5,000–10,000 × g. The lower centrifugal forces necessary to clarify Benzonase® nuclease treated cell extracts are more adaptable to largescale processes where generation of high centrifugal forces is impractical. Additionally, soluble protein supernatant yields are greater with Benzonase® nuclease treatment, because the precipitated materials form tight pellets upon centrifugation and less liquid is trapped than in the jelly-like pellets from untreated samples.
Digestion of nucleic acids
Figure 2 demonstrates the digestion of nucleic acids by Benzonase® nuclease from BugBuster® reagent extract of E. coli by agarose gel analysis and ethidium bromide staining. The Benzonase®nuclease titration series allows visual confirmation of progressive attack on nucleic acids and complete digestion as treatment levels increase from left to right. Digestion of the nucleic acids is the key to the effectiveness of Benzonase® reagent as a bioprocessing reagent for viscosity reduction.
Nucleic acid digestion by Benzonase® nuclease
Figure 2. Nucleic acid digestion by Benzonase® nuclease.The E. coli cells BL21(DE3) containing a pET construct were suspended in BugBuster® reagent (5 mL/g wet weight). Aliquots of the suspension were treated with the indicated amounts of Benzonase® nuclease for 30 min at room temperature. Samples were clarified by centrifugation and analyzed by agarose gel electrophoresis and ethidium bromide staining.
Table 2.IMAC purification results.
Analysis of BugBuster® reagent extracted
BugBuster® + Benzonase® reagents
Analysis of BugBuster® reagent extracted
BugBuster® reagent only
Figure 3. Analysis of BugBuster® + Benzonase® reagents extracted and IMAC purified His•Tag® protein. Samples were prepared and immobilized-metal affinity chromatography (IMAC) was performed as described in the text. The indicated samples were analyzed by SDS-PAGE on a 4–20% gradient gel stained with Coomassie blue.
Immobilized-metal affinity chromatography using His•Bind® Resins
Soluble proteins from the centrifuged extracts were applied by gravity flow to glass columns packed with His•Bind® Resin or Ni-NTA His•Bind® Resin (1.5 × 2.3 cm, ~4 mL bed volume). Unbound and weakly adsorbed proteins were washed from the columns with 10 column volumes of 20 mM Tris-HCl, pH 7.9 containing 5 mM imidazole and 0.5 M NaCl followed by 10 column volumes of 20 mM Tris-HCl, pH 7.9 containing 30 mM imidazole and 0.5 M NaCl. The His•Tag® fusion protein was eluted with five column volumes 20 mM TrisHCl, pH 7.9 containing 0.5 M imidazole and 0.5 M NaCl, and 1.5 mL fractions were collected. Fractions were analyzed and pooled according to densitometric scanning analysis of SDS-PAGE results and protein concentration determined by the BCA assay.12 Fractions 3–11 were pooled from each IMAC separation.
Results of the IMAC purification are shown in Table 2 and in Figure 3. Benzonase® nuclease treatment of the extract resulted in much higher column flow rates, higher product recovery, and greater product purity with both His•Bind® and Ni-NTA His•Bind® Resins, as compared to purification from extracts that had not been treated with Benzonase® nuclease.
Summary
Through the application of BugBuster® Protein Extraction Reagent and Benzonase® Nuclease, it is now possible to rapidly and efficiently extract and recover proteins expressed in E. coli without exposing them to the harsh conditions associated with ultrasonication and pressure disruption. Gentle release of intracellular proteins is achieved with BugBuster® reagent and extract viscosity reduced or eliminated by digestion of problematic nucleic acids with Benzonase® nuclease. The addition of Benzonase® nuclease for viscosity reduction and nucleic acid digestion provides several advantages over BugBuster® reagent treatment alone or other cellular disintegration techniques. Downstream processing advantages demonstrated independently and by these experiments include:
  • Decreased centrifugal forces required for extract clarification
  • Increased column flow rates and chromatographic performance13–16
  • Higher yields of protein/product13–16
  • Reduced processing time
  • Increased biomass content and fewerhomogenization cycles for extracts processed by expanded bed adsorption17
  • Degradation of nucleic acids fromrecombinant protein samples, enabling compliance with FDA guidelines for nucleic acid contamination9
Finally, Benzonase® nuclease digestion for nucleic removal is superior to traditional extraction or precipitation methods, which can reduce yield due to protein denaturation and coprecipitation. While DNase and RNase have also been used for viscosity reduction, Benzonase® nuclease has a higher specific activity, digests all forms of DNA and RNA, and has been genetically engineered and produced in bulk quantities specifically for biotechnological and biopharmaceutical applications.
Materials
Loading
References
1.
Holte SM, Givskov M, Riise E. 1992 . PRODUCTION IN ESCHERICHIA COLI OF EXTRACELLULAR SERRATIA SPP, HYDROLASES UP. 5173-418.
2.
1992 . BENZON PHARMA A/S Bacterial enzymes and method for their production EP0229866.
3.
Eaves GN, Jeffries CD. 1963. ISOLATION AND PROPERTIES OF AN EXOCELLULAR NUCLEASE OF SERRATIA MARCESCENS. 85(2):273-278. https://doi.org/10.1128/jb.85.2.273-278.1963
4.
Nestle M, Roberts W. 1969. An extracellular nuclease from Serratia marcescens I. Purification and some properties of the enzyme. Journal of Biological Chemistry . 244(19):5213-5218.
5.
Friedhoff P, Gimadutdinow O, Ruter T, Wende W, Urbanke C, Thole H, Pingoud A. 1994. A Procedure for Renaturation and Purification of the Extracellular Serratia marcescens Nuclease from Genetically Engineered Escherichia coli. Protein Expression and Purification. 5(1):37-43. https://doi.org/10.1006/prep.1994.1005
6.
Ball TK, Saurugger PN, Benedik MJ. 1987. The extracellular nuclease gene of Serratia marcescens and its secretion from Escherichia coli. Gene. 57(2-3):183-192. https://doi.org/10.1016/0378-1119(87)90121-1
7.
Ball TK, Suh Y, Benedik MJ. 1992. Disulfide bonds are required forSerratia marcescensnuclease activity. Nucl Acids Res. 20(19):4971-4974. https://doi.org/10.1093/nar/20.19.4971
8.
Miller MD, Tanner J, Alpaugh M, Benedik MJ, Krause KL. 1994. 2.1 Å structure of Serratia endonuclease suggests a mechanism for binding to double-stranded DNA. Nat Struct Mol Biol. 1(7):461-468. https://doi.org/10.1038/nsb0794-461
9.
1999. Merck KGaA Darmstadt, Germany Benzonase® Brochure, Code No.W 220911.
10.
Janning P, Schrader W, Linscheid M, Baillie TA. 1994. A new mass spectrometric approach to detect modifications in DNA. Rapid Commun. Mass Spectrom.. 8(12):1035-1040. https://doi.org/10.1002/rcm.1290081226
11.
Meiss G, Friedhoff P, Hahn M, Gimadutdinow O, Pingoud A. 1995. Sequence preferences in cleavage of dsDNA and ssDNA by the extracellular Serratia marcescens endonuclease. Biochemistry. 34(37):11979-11988. https://doi.org/10.1021/bi00037a040
12.
Smith P, Krohn R, Hermanson G, Mallia A, Gartner F, Provenzano M, Fujimoto E, Goeke N, Olson B, Klenk D. 1985. Measurement of protein using bicinchoninic acid. Analytical Biochemistry. 150(1):76-85. https://doi.org/10.1016/0003-2697(85)90442-7
13.
Huyghe BG, Liu X, Sutjipto S, Sugarman BJ, Horn MT, Shepard HM, Scandella CJ, Shabram P. 1995. Purification of a Type 5 Recombinant Adenovirus Encoding Human p53 by Column Chromatography. Human Gene Therapy. 6(11):1403-1416. https://doi.org/10.1089/hum.1995.6.11-1403
14.
Hagen A, Aboud R, DePhillips P, Oliver C, Orella C, Sitrin R. 1996 . Use of nuclease enzyme in the purification of VAQTA, a hepatitis A vaccine. . Biotechnology and applied biochemistry . 23(3):209-215.
15.
Hagen A, Oliver C, Sitrin R. 1996. Optimization of Poly(ethylene glycol) Precipitation of Hepatitis A Virus Used To Prepare VAQTA, a Highly Purified Inactivated Vaccine. Biotechnol. Prog.. 12(3):406-412. https://doi.org/10.1021/bp950081g
16.
COLOMBO S, D'AURIA S, FUSI P, ZECCA L, RAIA CA, TORTORA P. 1992. Purification and characterization of a thermostable carboxypeptidase from the extreme thermophilic archaebacterium Sulfolobus solfataricus. 206(2):349-357. https://doi.org/10.1111/j.1432-1033.1992.tb16934.x
17.
Barnfield Frej A, Hjorth R, Hammarström Å. 1994. Pilot scale recovery of recombinant annexin V from unclarifiedescherichia colihomogenate using expanded bed adsorption. Biotechnol. Bioeng.. 44(8):922-929. https://doi.org/10.1002/bit.260440808