Merck
HomeGel ElectrophoresisMolecular Weight Markers for Gel Filtration Chromatography
Molecular Weight Markers for Gel Filtration Chromatography
Gel filtration chromatography is a well-accepted method for determining the size and molecular weight of proteins. The molecular weight of a given protein may be determined by comparing its elution volume with those of known protein standards. An essentially linear relationship is obtained if the logarithms of the molecular weights of standard proteins are plotted against their respective ratios of elution volume to column void volume, the column void volume being the elution volume of a very large molecule such as Blue Dextran.
Examples of typical proteins used for size determination include:
ProteinApproximate Molecular Weight
Somatostatin1640
Aprotinin, Bovine Lung6,500
Cytochrome C, Horse Heart12,400
Carbonic Anhydrase, Bovine Erythrocytes29,000
Albumin, Bovine Serum66,000
Alchol Dehydrogenase, Yeast150,000
β-Amylase, Sweet Potato200,000
Apoferritin, Horse Spleen443,000
Thyroglobulin, Bovine669,000
Blue Dextran2,000,000
We supply these proteins as individual proteins, as well as molecular weight range kits. Many of the protein standards are also suitable for use in other chromatographic systems, including HPLC. Some buffer systems seem to alter the elution volumes of Albumin and Carbonic Anhydrase.
Examples of calibration curves:
Calibration curve typically obtained with specified proteins as run on Sephadex G-75-50. Calibration curve depicted should not be used to derive experimental results. Each laboratory must prepare its own calibration curves with its own system.
Calibration curve typically obtained with specified proteins as run on Sephadex G-75-50. Calibration curve depicted should not be used to derive experimental results. Each laboratory must prepare its own calibration curves with its own system.
	Calibration curve typically obtained with specified proteins as run on Sepharose 6B. Calibration curve depicted should not be used to derive experimental results. Each laboratory must prepare its own calibration curves with its own system.
Calibration curve typically obtained with specified proteins as run on Sepharose 6B. Calibration curve depicted should not be used to derive experimental results. Each laboratory must prepare its own calibration curves with its own system.
References:
1.
Whitaker, J.R . 1963. Determination of Molecular Weights of Proteins by Gel Filtration on Sephadex,. Anal Chem,1950.
2.
Andrews P. 1964. Estimation of the molecular weights of proteins by Sephadex gel-filtration. 91(2):222-233. https://doi.org/10.1042/bj0910222
3.
Marshall, J.J.. 1970. Chromatog. 53, 379