Pure & Simple Primers
Order Pure and Simple Primers, and you will be assured a quality start to all of your research applications with our ultra-high base coupling efficiency and optimized cartridge purification process.
Guaranteed Yield | 3 OD |
---|---|
Format | Shipped dry in 2 mL screw cap tubes |
Length | 12 - 35 bases |
Purification | Reverse-phase cartridge |
Bases | DNA (A, C, G or T) |
Modifications | None |
Backbone | Phosphodiester bond |
Documentation | All oligos are delivered with Quality Assurance documentation including yields, Tm (melting temperature), MW, and µg/OD |
Quality Control | Every oligo is quality checked by mass spectrometry |
Shipping Schedule | Ships within 24 - 48 hours |
Price & Ordering | Contact your local office for pricing and availability |
Sorry, an unexpected error has occurred
Network error: Failed to fetch