Determination of Water Content in Sugar Using Karl Fischer Titration
Product Group
Foodstuffs
General Information concerning the product group
Foodstuffs
Foodstuffs encompass the most diverse products. Depending on whether carbohydrate-rich, fatty or proteinrich substances are under investigation, different working techniques are favourable. Complex matrices that dissolve slowly, or where the water can only be slowly extracted, necessitate the use of solubilisers. In addition, titration under heating and the application of a homogeniser to accelerate water release are expedient.
Special Information concerning the sample and the methods
The dissolution of carbohydrates in methanolic KF solvents is often inadequate. For complete dissolution of the sample therefore the addition of formamide is necessary. However, the proportion of formamide should not exceed 50 %. Additionally, titration under warm conditions (50 °C) is favourable. With some types of sugar, however, heating leads to disintegration. If only the adherent moisture is of interest, the dissolution of the sample should be avoided. Due to the low water concentration the use of titrants with low factors are recommended. The coulometric method can only be applied after external solution of the sample in formamide and is therefore less recommended.
Titration one component system
Reagents:
Titrant
188002 Aquastar® - CombiTitrant 2 - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 2 mg water
Solvent
30 mL 188009 Aquastar® - CombiMethanol - Solvent for volumetric Karl Fischer titration with one component reagents, max. 0.01 % water
and
20 mL 109684 Formamide - as solubiliser
Titration Parameters:
Stirring time: 180 sec.
Temperature: 50 °C
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
2 g
Procedure:
The titration medium is first placed into the cell, heated up to 50 °C and titrated dry by means of the titrant. Then the sample is added from a weighing boat (exact sample weight determination by weighing of weighing boat before and after addition) and the titration is started. For complete dissolution of the sample a stirring time of at least 180 seconds is recommended. After two to three analyses it is recommended to change the KF solvent because the dissolution capacity of the solvent mixture decreases significantly.
Titration two component system
Reagents:
Titrant
188011 Aquastar® - Titrant 2 - Titrant for volumetric titration with two component reagents, 1 mL = approx. 2 mg water
Solvent
30 mL 188015 Aquastar® - Solvent - Solvent for volumetric titration with two component reagents
and
20 mL 109684 Formamide - as solubiliser
Titration Parameters:
Stirring time: 180 sec.
Temperature: 50 °C
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
2 g
Procedure:
The titration medium is first placed into the cell, heated up to 50 °C and titrated dry by means of the titrant. Then the sample is added from a weighing boat (exact sample weight determination by weighing of weighing boat before and after addition) and the titration is started. For complete dissolution of the sample a stirring time of at least 180 seconds is recommended. After two to three analyses it is recommended to change the KF solvent because the dissolution capacity of the solvent mixture decreases significantly.
Network error: Failed to fetch