Merck
HomeLarge Molecule HPLCHPLC Analysis of the Monoclonal Antibody (mAb) Erbitux (Cetuximab) on BIOshell A400 Protein C4
HPLC Analysis of the Monoclonal Antibody (mAb) Erbitux (Cetuximab) on BIOshell A400 Protein C4
HPLC Analysis of the Monoclonal Antibody (mAb) Erbitux (Cetuximab) on BIOshell™ A400 Protein C4
Conditions
ColumnBIOshell™ A400 Protein C4, 10 cm X 2.1 mm I.D., 3.4 µm particles (66825-U)
Column temp.90 °C
Mobile phase(A) 80:20, 0.1% TFA in water: 0.1% TFA in acetonitrile; (B) 20:80, 0.1% TFA in water: 0.1% TFA in acetonitrile
Gradient20 to 35%B in 10 min (or 5 min)
Flow rate0.2 mL/min
Pressure470 psi (or 940 psi)
Sample1 g/L mAb in 20 mM Tricine/KOH, pH 7
Injection0.4 µL
DetectorUV, 215 nm
Description
Analysis NoteAntibodies can exhibit considerable hydrophobicity, and so efficient reversed-phase elution of this intact mAb requires the use of high temperature. The top chromatogram illustrates that BIOshell Fused-Core® columns can be operated at high flow rate while maintaining good peak shape at a reasonable back pressure.
CategoriesAnalytical Chromatography, Proteins
Featured IndustryLife Science and Biopharma
Legal InformationBIOshell is a trademark of Sigma-Aldrich Co. LLC, Fused-Core is a registered trademark of Advanced Materials Technology, Inc.
SuitabilityApplication for HPLC
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch