GC Analysis of a Grain Fatty Acid Methyl Ester (FAME) Mix on SP®-2560
![GC Analysis of a Grain Fatty Acid Methyl Ester (FAME) Mix on SP®-2560 suitable for GC](/deepweb/assets/sigmaaldrich/product/images/392/302/196b3754-8893-483a-9d95-1b47eb4c71e9/800/196b3754-8893-483a-9d95-1b47eb4c71e9.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SP-2560, 100 m x 0.25 mm I.D., 0.20 μm film (24056)
oven
140 °C (5 min) to 240 °C at 4 °C/min
injection
1 μL split 100:1
detector
FID, 260 °C
carrier gas
helium, 20 cm/sec
inj. temp.
260 °C
Description
Legal Information
SP is a registered trademark of Sigma-Aldrich Co. LLC