HPLC Analysis of Simple Underivatized Sugars on apHera™ NH2
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
apHera NH2, 15 cm × 4.6 mm I.D., 5 μm particles (56401AST)
mobile phase
[A] water: [B] acetonitrile; (20:80, A:B)
flow rate
1.0 mL/min
column temp.
25 °C
detector
ELSD, 45 °C, 3.5 psi nitrogen
injection
10 μL
sample
500 μg/mL in 30:70, water: acetonitrile
Description
Legal Information
apHera is a trademark of Sigma-Aldrich Co. LLC