HPLC Analysis of Cisplatin and Monohydrated Cisplatin on SeQuant® ZIC-HILIC
CONDITIONS
column
Sequant ZIC-HILIC, PEEK, 150 x 2.1 mm, 5 μm, 200 Å (1.50454.0001)
mobile phase
[A] 25 mM ammonium formate, pH 6.5; [B] 1,4-dioxane; (20:80, A:B)
column temp.
ambient
flow rate
0.1 mL/min
detector
305 nm
injection
1 μL
Description
Analysis Note
diluent: mobile phase without buffer
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany