HPLC Analysis of AQC-Threonine Enantiomers on Astec® CHIROBIOTIC® T
![HPLC Analysis of AQC-Threonine Enantiomers on Astec® CHIROBIOTIC® T application for HPLC](/deepweb/assets/sigmaaldrich/product/images/210/269/745fa0e4-f945-41a9-8a74-7f33449024de/800/745fa0e4-f945-41a9-8a74-7f33449024de.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] 20 mM ammonium acetate, pH 4.1; [B] methanol; (90:10, A:B)
flow rate
0.9 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
1 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC