Merck
HPLC Analysis of a Monoclonal Antibody on Zenix® SEC-150, 30 cm X 4.6 mm I.D., 3 μm particles after Proteolysis with IdeS Protease
HPLC Analysis of a Monoclonal Antibody on Zenix® SEC-150, 30 cm X 4.6 mm I.D., 3 μm particles after Proteolysis with IdeS Protease application for HPLC
Materials
related product
Product No.
Description
Pricing
FabRICATOR®
for cleaving 2 mg IgG, 2000 U/vial
CONDITIONS
column
Zenix® SEC-150, 30 cm X 4.6 mm I.D., 3 μm particles (Z777018)
mobile phase
0.15 M potassium phosphate monobasic, pH 7.0 with potassium hydroxide
flow rate
0.35 mL/min
pressure
1450 psi (100 bar)
column temp.
ambient
detector
UV, 215 nm
injection
1 μL
sample
digested SO57 mAb, ~0.5 g/L
Description
Analysis Note
Proteolysis of antibodies can be useful for simplifying further analysis. This application demonstrates separation of the two fragments, F(ab′)2 and scFc, after digestion with IdeS protease
Legal Information
Zenix is a registered trademark of Sepax Technologies