Merck
HPLC Analysis of 25-Dihydroxyvitamin D2, 25-Hydroxyvitamin D3 and 3-epi-25-Hydroxyvitamin D3 on Ascentis® Express F5, 20 °C
HPLC Analysis of 25-Dihydroxyvitamin D2, 25-Hydroxyvitamin D3 and 3-epi-25-Hydroxyvitamin D3 on Ascentis® Express F5, 20 °C application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express F5, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
mobile phase component
Product No.
Description
Pricing
Ammonium formate
eluent additive for LC-MS, LiChropur, ≥99.0%
standard
Product No.
Description
Pricing
3-epi-25-Hydroxyvitamin D3
98% (CP)
25-Hydroxyvitamin D2 solution
50 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
25-Hydroxyvitamin D3 solution
100 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
CONDITIONS
column
Ascentis Express F5, 10 cm x 2.1 mm I.D., 2.7 μm particles (53569-U)
mobile phase
[A] 7 mM ammonium formate; [B] methanol (25:75)
flow rate
0.4 mL/min
pressure
5221 psi (245 bar)
column temp.
20 °C
detector
(UV, 265 nm, ESI(+), m/z 100 - 1000)
injection
1 μL
sample
300 mg/L in 25:75, water:methanol
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany