Merck
GC Analysis of Phenols on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
GC Analysis of Phenols on SLB®-5ms after SPME using 60 μm Carbowax® Fiber application for SPME, suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.50 μm
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating
needle size 23 ga, df 60 μm(PEG, for use with autosampler
CONDITIONS
sample/matrix
Phenols at 25 ppb in 0.1M phosphate buffer, pH 2, with 25% NaCl, 9 mL in 10 mL screw cap vial
SPME fiber
60 μm Carbowax (PEG) (57354-U)
extraction
30 min direct immersion with agitation using CombiPal autosampler
desorption process
(30 min. direct immersion with agitation using CombiPal autosampler)
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.5μm film
oven
50°C(2.0 min) to 160°C @ 10°C/min to 280°C @ 20°C/min
inj. temp.
Splitless/split with 0.75mm ID liner and Merlin Microseal
detector
ITMS m/z=45-275 @ 0.7 μsec per scan, transfer line 310°C
carrier gas
Helium, 1mL/min constant flow, 8.7 psi
Description
Legal Information
Carbowax is a registered trademark of Union Carbide
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany