Merck
GC Analysis of Cold-Pressed Mandarin Essential Oil on SLB®-5ms
GC Analysis of Cold-Pressed Mandarin Essential Oil on SLB®-5ms suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
50°C at 3°C/min to 250°C, held 10 min, at 10°C/min to 270°C, held 3 min
detector
FID, 280 °C
carrier gas
He at 30.1 cm/s and pressure of 99.8 kPa
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany