GC Analysis of a 12-Component Chlorinated Solvents Mix on SLB®-IL60, Comparison to a Polyethylene Glycol (PEG) Column
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
PEG 2, 30 m x 0.25 mm I.D., 0.25 μm
column
SLB-IL60, 30 m x 0.25 mm I.D., 0.20 μm (29505-U)
oven
40 °C (4 min), 8 °C/min to 200 °C (5 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 cm/sec
injection
1 μL 100:1 split
liner
4 mm I.D., split/splitless type, single taper wool packed FocusLiner™ design
sample
12-component chlorinated solvent mix, each analyte at 0.2% (v/v) in pentane
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany