Merck
  • Inhibition of Intermediate-Conductance Calcium-Activated K Channel (KCa3.1) and Fibroblast Mitogenesis by α-Linolenic Acid and Alterations of Channel Expression in the Lysosomal Storage Disorders, Fabry Disease, and Niemann Pick C.
Inhibition of Intermediate-Conductance Calcium-Activated K Channel (KCa3.1) and Fibroblast Mitogenesis by α-Linolenic Acid and Alterations of Channel Expression in the Lysosomal Storage Disorders, Fabry Disease, and Niemann Pick C.
Frontiers in physiology (2017-02-16)
Aida Oliván-Viguera, Javier Lozano-Gerona, Laura López de Frutos, Jorge J Cebolla, Pilar Irún, Edgar Abarca-Lachen, Ana J García-Malinis, Ángel Luis García-Otín, Yolanda Gilaberte, Pilar Giraldo, Ralf Köhler
ABSTRACT
The calcium/calmodulin-gated KCa3.1 channel regulates normal and abnormal mitogenesis by controlling K
Sigma-Aldrich
SKA-121, ≥98% (HPLC)