Merck
  • Preclinical evaluation of the Aurora kinase inhibitors AMG 900, AZD1152-HQPA, and MK-5108 on SW-872 and 93T449 human liposarcoma cells.
Preclinical evaluation of the Aurora kinase inhibitors AMG 900, AZD1152-HQPA, and MK-5108 on SW-872 and 93T449 human liposarcoma cells.
In vitro cellular & developmental biology. Animal (2017-12-03)
Sandhya Noronha, Lauren A C Alt, Taylor E Scimeca, Omran Zarou, Justyna Obrzut, Brian Zanotti, Elizabeth A Hayward, Akhil Pillai, Shubha Mathur, Joseph Rojas, Ribhi Salamah, Nalini Chandar, Michael J Fay
ABSTRACT
Liposarcoma is a malignant soft tissue tumor that originates from adipose tissue and is one of the most frequently diagnosed soft tissue sarcomas in humans. There is great interest in identifying novel chemotherapeutic options for treating liposarcoma based upon molecular alterations in the cancer cells. The Aurora kinases have been identified as promising chemotherapeutic targets based on their altered expression in many human cancers and cellular roles in mitosis and cytokinesis. In this study, we investigated the effects of an Aurora kinase A inhibitor (MK-5108), an Aurora kinase B inhibitor (AZD1152-HQPA), and a pan-Aurora kinase inhibitor (AMG 900) on undifferentiated SW-872 and well-differentiated 93T449 human liposarcoma cells. Treatment of the SW-872 and 93T449 cells with MK-5108 (0-1000 nM), AZD1152-HQPA (0-1000 nM), and AMG 900 (0-1000 nM) for 72 h resulted in a dose-dependent decrease in the total viable cell number. Based upon the EC
Sigma-Aldrich
AZD1152-HQPA, ≥98% (HPLC)