Merck
  • Organophosphate pesticide in agricultural soils from the Yangtze River Delta of China: concentration, distribution, and risk assessment.
Organophosphate pesticide in agricultural soils from the Yangtze River Delta of China: concentration, distribution, and risk assessment.
Environmental science and pollution research international (2016-10-01)
Lili Pan, Jianteng Sun, Zhiheng Li, Yu Zhan, Shen Xu, Lizhong Zhu
ABSTRACT
Organophosphorus pesticides (OPPs) are used worldwide and pose great risks to human health. However, information on their presence in agricultural soils at regional scale and the associated risks is limited. In this study, an extensive investigation on agricultural soils was conducted throughout the Yangtze River Delta (YRD) of China to reveal the status of OPP pollution. The total concentrations of the nine OPPs ranged from <3.0 to 521 ng g
Product Description
Supelco
Sulfotep, PESTANAL®, analytical standard