Merck
Hierarchical Porous Carbon Spheres for High-Performance Na-O
Advanced materials (Deerfield Beach, Fla.) (2017-04-05)
Bing Sun, Katja Kretschmer, Xiuqiang Xie, Paul Munroe, Zhangquan Peng, Guoxiu Wang
ABSTRACT
As a new family member of room-temperature aprotic metal-O
Sigma-Aldrich
Sodium trifluoromethanesulfonate, 98%
Pricing and availability is not currently available.