Merck
  • Gas chromatographic retention behavior of polycyclic aromatic hydrocarbons (PAHs) and alkyl-substituted PAHs on two stationary phases of different selectivity.
Gas chromatographic retention behavior of polycyclic aromatic hydrocarbons (PAHs) and alkyl-substituted PAHs on two stationary phases of different selectivity.
Analytical and bioanalytical chemistry (2017-11-10)
Federica Nalin, Lane C Sander, Walter B Wilson, Stephen A Wise
ABSTRACT
Retention indices (I) for 45 polycyclic aromatic hydrocarbons (PAHs) and 63 methyl-substituted PAHs were determined by gas chromatography - mass spectrometry (GC-MS) using two different stationary phases: a Rxi-PAH phase (a "higher phenyl-content stationary phase") and a 50% (mole fraction) liquid crystalline dimethylpolysiloxane phase. Retention data were obtained for parent PAHs from molecular mass (MM) 128 g/mol (naphthalene) to 328 g/mol (benzo[c]picene) and for 12 sets of methyl-PAHs (methylfluorenes, methylanthracenes, methylphenanthrenes, methylfluoranthenes, methylpyrenes, methylbenz[a]anthracenes, methylbenzo[c]phenanthrenes, methylchrysenes, methyltriphenylenes, methylbenzo[a]pyrenes, methylperylenes, and methylpicenes). Molecular shape descriptors such as length-to-breath ratio (L/B) and thickness (T) were determined for all the PAHs studied. Correlation between I and L/B ratio was evaluated for both stationary phases with a better correlation observed for the 50% liquid crystalline phase (correlation coefficients ranging from 0.22 to 1.00). Graphical Abstract GC separation of six methylchrysene isomers (m/z 242) on two different stationary phases: 50 % phenyl-like methylpolysiloxane phase and 50 % liquid crystalline phase. Retention indices (I) are plotted as a function of L/B for both phases. The data marker numbers identify each isomer based on methyl-substitution position.