Merck
  • The BEACH Protein LRBA Promotes the Localization of the Heterotrimeric G-protein G
The BEACH Protein LRBA Promotes the Localization of the Heterotrimeric G-protein G
Scientific reports (2017-08-18)
Stefan Kurtenbach, Andreas Gießl, Siv Strömberg, Jan Kremers, Jenny Atorf, Sebastian Rasche, Eva M Neuhaus, Denis Hervé, Johann Helmut Brandstätter, Esther Asan, Hanns Hatt, Manfred W Kilimann
ABSTRACT
BEACH domain proteins are involved in membrane protein traffic and human diseases, but their molecular mechanisms are not understood. The BEACH protein LRBA has been implicated in immune response and cell proliferation, and human LRBA mutations cause severe immune deficiency. Here, we report a first functional and molecular phenotype outside the immune system of LRBA-knockout mice: compromised olfaction, manifesting in reduced electro-olfactogram response amplitude, impaired food-finding efficiency, and smaller olfactory bulbs. LRBA is prominently expressed in olfactory and vomeronasal chemosensory neurons of wild-type mice. Olfactory impairment in the LRBA-KO is explained by markedly reduced concentrations (20-40% of wild-type levels) of all three subunits α
Sigma-Aldrich
Anti-MAGI-2 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Radixin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Pan Cadherin antibody produced in mouse, clone CH-19, ascites fluid