Merck
  • Pd/P(t-Bu)(3): a mild and general catalyst for Stille reactions of aryl chlorides and aryl bromides.
Pd/P(t-Bu)(3): a mild and general catalyst for Stille reactions of aryl chlorides and aryl bromides.
Journal of the American Chemical Society (2002-05-30)
Adam F Littke, Lothar Schwarz, Gregory C Fu
ABSTRACT
Pd/P(t-Bu)(3) serves as an unusually reactive catalyst for Stille reactions of aryl chlorides and bromides, providing solutions to a number of long-standing challenges. An unprecedented array of aryl chlorides can be cross-coupled with a range of organotin reagents, including SnBu(4). Very hindered biaryls (e.g., tetra-ortho-substituted) can be synthesized, and aryl chlorides can be coupled in the presence of aryl triflates. The method is user-friendly, since a commercially available complex, Pd(P(t-Bu)(3))(2), is effective. Pd/P(t-Bu)(3) also functions as an active catalyst for Stille reactions of aryl bromides, furnishing the first general method for room-temperature cross-couplings.
Sigma-Aldrich
Bis(tri-tert-butylphosphine)palladium(0)
Sigma-Aldrich
P(t-Bu)3 Pd G2