Merck
  • Vardenafil reduces macrophage pro-inflammatory overresponses in cystic fibrosis through PDE5- and CFTR-dependent mechanisms.
Vardenafil reduces macrophage pro-inflammatory overresponses in cystic fibrosis through PDE5- and CFTR-dependent mechanisms.
Clinical science (London, England : 1979) (2017-02-16)
Sabrina Noel, Nadtha Panin, Mathilde Beka, Barbara Dhooghe, François Huaux, Teresinha Leal
ABSTRACT
Chronic inflammation that progressively disrupts the lung tissue is a hallmark of cystic fibrosis (CF). In mice, vardenafil, an inhibitor of phosphodiesterase type 5 (PDE5), restores transepithelial ion transport and corrects mislocalization of the most common CF mutation, F508del-CFTR. It also reduces lung pro-inflammatory responses in mice and in patients with CF. To test the hypothesis that macrophages are target effector cells of the immunomo-dulatory effect of vardenafil, we isolated lung macrophages from mice homozygous for the F508del mutation or invalidated for the
Sigma-Aldrich
Vardenafil hydrochloride trihydrate, ≥98% (HPLC)