Merck
  • The efficacy of rituximab plus Hyper-CVAD regimen in mantle cell lymphoma is independent of FCgammaRIIIa and FCgammaRIIa polymorphisms.
The efficacy of rituximab plus Hyper-CVAD regimen in mantle cell lymphoma is independent of FCgammaRIIIa and FCgammaRIIa polymorphisms.
Journal of chemotherapy (Florence, Italy) (2007-06-28)
S Galimberti, G A Palumbo, F Caracciolo, E Benedetti, M Pelosini, S Brizzi, E Ciabatti, R Fazzi, C Stelitano, G Quintana, E Conte, D Tibullo, F Di Raimondo, M Petrini
ABSTRACT
Mantle cell lymphoma (MCL) accounts for 3-10% of all non-Hodgkin's lymphomas, with median overall survival not exceeding 3-4 years. Rituximab in combination with the Hyper-CVAD regimen appears the most promising regimen; thus, we adopted it as a first-line treatment strategy in a series of 24 patients. In addition to evaluation of clinical success of the regimen, we investigated a possible role of polymorphism in IgG Fc receptors, FCgammaRIIIa and FCgammaRIIa. The frequencies of FCgammaRIIIa-158 were as follows: V/V=4/24 (17%); V/F=16/24 (66%); F/F=4/24 (17%). Those of the FCgammaRIIa-131 polymorphism were H/H=11/24 (46%), H/R=9/24 (37%), R/R=4/24 (17%). The overall response rate was 62.5%, with 33% of complete responses (CRs) after four cycles of R-Hyper-CVAD. Two-year progression-free survival (PFS) was 78% for 158V/V patients vs 75% for cases carrying phenylalanine (p=0.88). When the FCgammaRIIa polymorphism was assessed, the 2-year PFS was 82% for 131H/H patients vs 75% for those carrying arginine (p=0.26). Eighty-three percent of cases achieved Polymerase Chain Reaction (PCR)-negativity: the progression rate was significantly influenced by the minimal residual disease clearance, with 12% progression in the subgroup of PCR-negative cases versus 67% progression in PCR-positive cases (p=0.008). The achievement of PCRnegativity was not significantly influenced by FCgammaR polymorphisms. Results confirm that rituximab plus Hyper-CVAD is an effective regimen for the induction of prolonged remission in patients with aggressive MCL and suggest that rituximab efficacy is independent of the FCgammaR polymorphisms.