Merck
  • Exogenous Glycine Nitrogen Enhances Accumulation of Glycosylated Flavonoids and Antioxidant Activity in Lettuce (
Exogenous Glycine Nitrogen Enhances Accumulation of Glycosylated Flavonoids and Antioxidant Activity in Lettuce (
Frontiers in plant science (2018-01-13)
Xiao Yang, Xiaoxian Cui, Li Zhao, Doudou Guo, Lei Feng, Shiwei Wei, Chao Zhao, Danfeng Huang
ABSTRACT
Glycine, the simplest amino acid in nature and one of the most abundant free amino acids in soil, is regarded as a model nutrient in organic nitrogen studies. To date, many studies have focused on the uptake, metabolism and distribution of organic nitrogen in plants, but few have investigated the nutritional performance of plants supplied with organic nitrogen. Lettuce (
Sigma-Aldrich
Trifluoroacetamide, 97%
Pricing and availability is not currently available.