Merck
  • Differential Anticancer Activity of Pterostilbene Against Three Subtypes of Human Breast Cancer Cells.
Differential Anticancer Activity of Pterostilbene Against Three Subtypes of Human Breast Cancer Cells.
Anticancer research (2017-10-25)
Rei Wakimoto, Misaki Ono, Mikako Takeshima, Takako Higuchi, Shuji Nakano
ABSTRACT
Although pterostilbene, a natural analog of resveratrol, has potent antitumor activity against several human cancer types, the possible inhibitory mechanisms against subtypes of human breast cancer with different hormone receptor and human epidermal growth factor receptor 2 (HER2) status remain unknown. We investigated the anticancer activity of pterostilbene using three subtypes of breast cancer cell lines. Pterostilbene treatment exhibited a dose-dependent antiproliferative activity, with the greatest growth inhibition observed in triple-negative MDA-MB-468 cells. Although pterostilbene arrested cell-cycle progression at the G
Sigma-Aldrich
Pterostilbene, ≥97% (HPLC), solid
Pricing and availability is not currently available.