Merck
  • Comprehensive Quality Assessment Based Specific Chemical Profiles for Geographic and Tissue Variation in
Comprehensive Quality Assessment Based Specific Chemical Profiles for Geographic and Tissue Variation in
Frontiers in chemistry (2018-01-10)
Jie Li, Ji Zhang, Yan-Li Zhao, Heng-Yu Huang, Yuan-Zhong Wang
ABSTRACT
Roots, stems, leaves, and flowers of Longdan (
Sigma-Aldrich
Loganic acid, ≥95% (LC/MS-ELSD)