Merck
  • Construction of proton exchange membranes under ultrasonic irradiation based on novel fluorine functionalizing sulfonated polybenzimidazole/cellulose/silica bionanocomposite.
Construction of proton exchange membranes under ultrasonic irradiation based on novel fluorine functionalizing sulfonated polybenzimidazole/cellulose/silica bionanocomposite.
Ultrasonics sonochemistry (2017-11-16)
Sheida Esmaielzadeh, Hashem Ahmadizadegan
ABSTRACT
Novel sulfonated polybenzimidazole (s-PBI)/cellulose/silica bionanocomposite membranes were prepared from fluorine-containing s-PBI copolymer with a cellulose/silica precursor and a bonding agent. The introduction of the bonding agent results in the reinforcing interfacial interaction between s-PBI chains and the cellulose/silica nanoparticles. Commercially available silica nanoparticles were modified with biodegradable nanocellolose through ultrasonic irradiation technique. Transmission electron microscopy (TEM) analyses showed that the cellulose/silica composites were well dispersed in the s-PBI matrix on a nanometer scale. The mechanical properties and the methanol barrier ability of the s-PBI films were improved by the addition of cellulose/silica. The modulus of the s-PBI/10 wt% cellulose/silica nanocomposite membranes had a 45% increase compared to the pure s-PBI films, and the methanol permeability decreased by 62% with respect to the pure s-PBI membranes. The conductivities of the s-PBI/cellulose/silica nanocomposites were slightly lower than the pure s-PBI. The antibacterial activity of (s-PBI)/cellulose/silica was investigated against Gram-positive bacteria, ie, Staphylococcus aureus and methicillin-resistant S. aureus and Gram-negative bacteria, ie, Escherichia coli, E. coli O157:H7 and Pseudomonas aeruginosa by the disc diffusion method using Mueller Hinton agar at different sizes of cellulose/silica. All of the synthesized (s-PBI)/cellulose/silica were found to have high antibacterial activity.
Sigma-Aldrich
4,4′-(Hexafluoroisopropylidene)bis(benzoic acid), 98%