Merck
Broad-spectrum antiviral agents: secreted phospholipase A
Scientific reports (2017-11-23)
Ming Chen, Chie Aoki-Utsubo, Masanori Kameoka, Lin Deng, Yutaka Terada, Wataru Kamitani, Kei Sato, Yoshio Koyanagi, Makoto Hijikata, Keiko Shindo, Takeshi Noda, Michinori Kohara, Hak Hotta
ABSTRACT
Hepatitis C virus (HCV), dengue virus (DENV) and Japanese encephalitis virus (JEV) belong to the family Flaviviridae. Their viral particles have the envelope composed of viral proteins and a lipid bilayer acquired from budding through the endoplasmic reticulum (ER). The phospholipid content of the ER membrane differs from that of the plasma membrane (PM). The phospholipase A
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Phospholipase A2 from bovine pancreas, lyophilized powder, ≥20 units/mg protein