- Arginase 1 deletion in myeloid cells affects the inflammatory response in allergic asthma, but not lung mechanics, in female mice.
Arginase 1 deletion in myeloid cells affects the inflammatory response in allergic asthma, but not lung mechanics, in female mice.
(Over-)expression of arginase may limit local availability of arginine for nitric oxide synthesis. We investigated the significance of arginase1 (ARG1) for the development of airway hyperresponsiveness (AHR) and lung inflammation in female mice with ovalbumin (OVA)-induced allergic asthma. Arg1 was ablated in the lung by crossing Arg1 Arg1 ablation had no influence on the development of OVA-induced AHR, but attenuated OVA-induced increases in expression of Arg2 and Nos2, Slc7a1, Slc7a2, and Slc7a7 (arginine transporters), Il4, Il5 and Il13 (T Complete ablation of Arg1 in the lung affects mRNA abundance of arginine-transporting and -metabolizing genes, and pro-inflammatory genes, but not methacholine responsiveness or accumulation of inflammatory cells.