Merck
  • Microbiome symbionts and diet diversity incur costs on the immune system of insect larvae.
Microbiome symbionts and diet diversity incur costs on the immune system of insect larvae.
The Journal of experimental biology (2017-09-25)
Indrikis A Krams, Sanita Kecko, Priit Jõers, Giedrius Trakimas, Didzis Elferts, Ronalds Krams, Severi Luoto, Markus J Rantala, Inna Inashkina, Dita Gudrā, Dāvids Fridmanis, Jorge Contreras-Garduño, Lelde Grantiņa-Ieviņa, Tatjana Krama
ABSTRACT
Communities of symbiotic microorganisms that colonize the gastrointestinal tract play an important role in food digestion and protection against opportunistic microbes. Diet diversity increases the number of symbionts in the intestines, a benefit that is considered to impose no cost for the host organism. However, less is known about the possible immunological investments that hosts have to make in order to control the infections caused by symbiont populations that increase because of diet diversity. Using taxonomical composition analysis of the
Sigma-Aldrich
Kanamycin solution from Streptomyces kanamyceticus, 50 - 60 mg/mL in 0.9% NaCl, BioReagent, liquid, 0.1 μm filtered, suitable for cell culture
Sigma-Aldrich
Gentamicin solution, 50 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ampicillin sodium salt, Ready Made Solution, 100 mg/mL, 0.2 μm filtered