- Antiproliferative activity of
Antiproliferative activity of
Pharmacognosy magazine (2017-08-16)
J Bello-Martínez, M Jiménez-Estrada, J L Rosas-Acevedo, L P Avila-Caballero, M Vidal-Gutierrez, C Patiño-Morales, E Ortiz-Sánchez, R E Robles-Zepeda
PMID28808394
ABSTRACT
The aim of this study was to isolate the compounds responsible for antiproliferative activity of A bioassay-guided fractionation of ethanol extract of The flavonoid brazilin (1) was isolated from the heartwood of Brazilin: (6a The flavonoid brazilin was isolated from the heartwood of