- Conductive Polymer Waving in Liquid Nitrogen.
Conductive Polymer Waving in Liquid Nitrogen.
The poor mechanical properties and processability of pristine heterocyclic conductive polymers represent the most notable scientific and technological challenges that have greatly limited the application of these polymers. We report a soft and mechanically processable free-standing pristine polypyrrole (PPy) membrane (PPy-N) that is as soft in liquid nitrogen (-196 °C) as it is at room temperature, despite a glass transition temperature (T