Merck
Self-Assembly of Cyclohelicate [M
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-06-09)
Ross W Hogue, Sébastien Dhers, Ryan M Hellyer, Jingwei Luo, Garry S Hanan, David S Larsen, Anna L Garden, Sally Brooker
ABSTRACT
Self-assembly of 1:1:2 M
Sigma-Aldrich
Manganese(IV) oxide, activated, ~85%, <10 μm
Pricing and availability is not currently available.