Merck
  • Preparation of Ti species coating hydrotalcite by chemical vapor deposition for photodegradation of azo dye.
Preparation of Ti species coating hydrotalcite by chemical vapor deposition for photodegradation of azo dye.
Journal of environmental sciences (China) (2017-10-17)
Gaofei Xiao, HongYan Zeng, Sheng Xu, ChaoRong Chen, Quan Zhao, XiaoJun Liu
ABSTRACT
TiO