Merck
  • Use of tellurite for the selection of verocytotoxigenic Escherichia coli O157.
Use of tellurite for the selection of verocytotoxigenic Escherichia coli O157.
Journal of medical microbiology (1993-08-01)
P M Zadik, P A Chapman, C A Siddons
ABSTRACT
Potassium tellurite was assessed for the selection of verocytotoxigenic (VT+) Escherichia coli O157. MICs were higher for VT+ E. coli O157 than for other strains of E. coli and for Aeromonas spp. MacConkey medium containing sorbitol, tellurite and cefixime (TC-SMAC) permitted the growth of VT+ E. coli O157 and Shigella sonnei but partially or completely inhibited the growth of 67% of other strains of E. coli and all or most strains of other sorbitol-non-fermenting species tested. Of 391 rectal swabs from cattle screened on TC-SMAC medium, 26 yielded isolates of VT+ E. coli O157 whereas sorbitol-MacConkey medium with cefixime and rhamnose yielded only nine isolates. Inclusion of potassium tellurite in sorbitol-MacConkey agar markedly increased the rate of isolation of VT+ E. coli O157 from cattle rectal swabs and may do so for other types of specimen.
Product Description
Supelco
Cefixime trihydrate, suitable for microbiology, for ion-selective electrodes, ≥98.0% (HPLC)